Assay Details
| Gene Symbol: Cpr47Eg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136817.4 | NP_610661.1 | 2 | 247 | 78 | |
| GenBank mRNA | AY071014.1 | - | 2 | 243 | 78 | |
| KX531827.1 | - | 2 | 208 | 78 | ||
More Information
Set Membership: |
|

Cpr47Eg
Genomic Map