Assay Details
| Gene Symbol: Cpr56F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137626.3 | NP_611470.1 | 2 | 1031 | 93 | |
| GenBank mRNA | AY071231.1 | - | 2 | 1028 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr56F| Gene Symbol: Cpr56F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137626.3 | NP_611470.1 | 2 | 1031 | 93 | |
| GenBank mRNA | AY071231.1 | - | 2 | 1028 | 93 | |
Set Membership: |
|