Assay Details
| Gene Symbol: Nf-YA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140056.3 | NP_648313.1 | 2 | 1859 | 170 | |
| GenBank mRNA | AY118913.1 | - | 2 | 1821 | 170 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nf-YA| Gene Symbol: Nf-YA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140056.3 | NP_648313.1 | 2 | 1859 | 170 | |
| GenBank mRNA | AY118913.1 | - | 2 | 1821 | 170 | |
Set Membership: |
|