Assay Details
| Gene Symbol: Su(Tpl) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170006.2 | NP_001163477.1 | 5 | 4215 | 67 | |
| NM_001300193.1 | NP_001287122.1 | 5 | 4264 | 67 | ||
| NM_140898.5 | NP_649155.3 | 5 | 4220 | 67 | ||
| GenBank mRNA | BT050407.1 | - | 5 | 4185 | 67 | |
| BT058015.1 | - | 5 | 4190 | 67 | ||
More Information
Set Membership: |
|

Su(Tpl)
Genomic Map