Assay Details
| Gene Symbol: Cpr78Cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141043.3 | NP_649300.1 | 2 | 244 | 78 | |
| GenBank mRNA | AY113448.1 | - | 2 | 236 | 78 | |
| KX531918.1 | - | 1 | 203 | 78 | ||
More Information
Set Membership: |
|

Cpr78Cc
Genomic Map