Assay Details
| Gene Symbol: Cpr78E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141088.2 | NP_649345.1 | 2 | 398 | 91 | |
| GenBank mRNA | AY119036.1 | - | 2 | 398 | 91 | |
| KX531817.1 | - | 1 | 374 | 91 | ||
More Information
Set Membership: |
|

Cpr78E
Genomic Map