Assay Details
| Gene Symbol: GluClalpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275811.2 | NP_001262740.1 | 12 | 5397 | 66 | |
| NM_001300480.1 | NP_001287409.1 | 12 | 5391 | 66 | ||
| NM_142570.4 | NP_650827.3 | 12 | 5394 | 66 | ||
| NM_169873.4 | NP_732447.2 | 12 | 5394 | 66 | ||
More Information
Set Membership: |
|

GluClalpha
Genomic Map