Assay Details
| Gene Symbol: TBCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164570.2 | NP_722959.1 | 2 | 792 | 55 | |
| NM_164571.2 | NP_722960.1 | 2 | 936 | 55 | ||
| GenBank mRNA | AY058646.1 | - | 2 | 774 | 55 | |
More Information
Set Membership: |
|

TBCC
Genomic Map