Assay Details
| Gene Symbol: Rab30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135250.4 | NP_609094.1 | 2 | 1353 | 76 | |
| NM_164728.3 | NP_723241.1 | 2 | 1364 | 76 | ||
| NM_164729.3 | NP_723242.1 | 2 | 1438 | 76 | ||
| GenBank mRNA | AY071080.1 | - | 2 | 1350 | 76 | |
More Information
Set Membership: |
|

Rab30
Genomic Map