Assay Details
| Gene Symbol: Ggamma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165631.2 | NP_724718.1 | 2 | 1116 | 119 | |
| GenBank mRNA | M85042.1 | - | 2 | 1101 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggamma1| Gene Symbol: Ggamma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165631.2 | NP_724718.1 | 2 | 1116 | 119 | |
| GenBank mRNA | M85042.1 | - | 2 | 1101 | 119 | |
Set Membership: |
|