Assay Details
| Gene Symbol: Ggamma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078938.3 | NP_523662.1 | 2 | 570 | 127 | |
| NM_165631.2 | NP_724718.1 | 2 | 632 | 127 | ||
| NM_165632.2 | NP_724719.1 | 2 | 823 | 127 | ||
| GenBank mRNA | AY118489.1 | - | 2 | 803 | 127 | |
| M85042.1 | - | 2 | 617 | 127 | ||
More Information
Set Membership: |
|

Ggamma1
Genomic Map