Assay Details
| Gene Symbol: SRPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382195.1 | NP_001369087.1 | 5 | 4370 | 126 | |
| NM_137190.4 | NP_611034.2 | 4 | 4365 | 126 | ||
| NM_166094.1 | NP_725459.1 | 3 | 3732 | 126 | ||
More Information
Set Membership: |
|

SRPK
Genomic Map