Assay Details
| Gene Symbol: RpS14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080145.5 | NP_524884.1 | 3 | 549 | 94 | |
| NM_167137.2 | NP_727218.1 | 2 | 783 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RpS14a| Gene Symbol: RpS14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080145.5 | NP_524884.1 | 3 | 549 | 94 | |
| NM_167137.2 | NP_727218.1 | 2 | 783 | 94 | ||
Set Membership: |
|