Assay Details
| Gene Symbol: CG33181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103438.2 | NP_001096908.1 | 2 | 429 | 153 | |
| NM_001103441.2 | NP_001096911.1 | 2 | 518 | 153 | ||
| NM_001103442.2 | NP_001096912.1 | 2 | 637 | 153 | ||
| NM_001272405.1 | NP_001259334.1 | 3 | 512 | 153 | ||
| NM_001272406.1 | NP_001259335.1 | 3 | 403 | 153 | ||
| GenBank mRNA | BT120117.1 | - | 2 | 391 | 153 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT120117.1, NM_001103438.2, NM_001103441.2, NM_001103442.2, NM_001272405.1, NM_001272406.1 transcripts.
Set Membership: |
|

CG33181
Genomic Map