Assay Details
| Gene Symbol: dsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078563.3 | NP_511118.2 | 1 | 2735 | 123 | |
| GenBank mRNA | BT023781.1 | - | 1 | 2713 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dsh| Gene Symbol: dsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078563.3 | NP_511118.2 | 1 | 2735 | 123 | |
| GenBank mRNA | BT023781.1 | - | 1 | 2713 | 123 | |
Set Membership: |
|