Assay Details
| Gene Symbol: Jafrac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298273.1 | NP_001285202.1 | 2 | 600 | 57 | |
| NM_001298274.1 | NP_001285203.1 | 2 | 629 | 57 | ||
| NM_001298275.1 | NP_001285204.1 | 1 | 525 | 57 | ||
| NM_058162.3 | NP_477510.1 | 2 | 559 | 57 | ||
| NM_167359.2 | NP_727689.1 | 2 | 563 | 57 | ||
| GenBank mRNA | AF167098.1 | - | 2 | 477 | 57 | |
| AF321615.1 | - | 2 | 543 | 57 | ||
| AF321616.1 | - | 2 | 556 | 57 | ||
| AY070534.1 | - | 2 | 539 | 57 | ||
| BT014630.1 | - | 922 | 57 | |||
More Information
Set Membership: |
|

Jafrac1
Genomic Map