Assay Details
| Gene Symbol: sty | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168031.2 | NP_728881.1 | 2 | 3454 | 110 | |
| GenBank mRNA | BT150269.1 | - | 2 | 3455 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sty| Gene Symbol: sty | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168031.2 | NP_728881.1 | 2 | 3454 | 110 | |
| GenBank mRNA | BT150269.1 | - | 2 | 3455 | 110 | |
Set Membership: |
|