Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274987.1 | NP_001261916.1 | 2 | 1157 | 69 | |
| NM_001274988.1 | NP_001261917.1 | 3 | 1707 | 69 | ||
| NM_001274989.1 | NP_001261918.1 | 2 | 1636 | 69 | ||
| NM_079377.3 | NP_524101.2 | 2 | 1421 | 69 | ||
| NM_168644.2 | NP_730097.1 | 2 | 1273 | 69 | ||
| NM_168645.2 | NP_730098.1 | 2 | 1223 | 69 | ||
| GenBank mRNA | BT031197.1 | - | 2 | 1255 | 69 | |
More Information
Set Membership: |
|

Diap1
Genomic Map