Assay Details
| Gene Symbol: chb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079912.4 | NP_524651.2 | 2 | 3179 | 68 | |
| NM_168882.2 | NP_730596.1 | 2 | 2624 | 68 | ||
| NM_168883.1 | NP_730597.1 | 2 | 2513 | 68 | ||
| GenBank mRNA | AF195498.1 | - | 2 | 3165 | 68 | |
| AF250842.1 | - | 2 | 3164 | 68 | ||
| AY069579.1 | - | 2 | 3160 | 68 | ||
More Information
Set Membership: |
|

chb
Genomic Map