Assay Details
| Gene Symbol: hb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169233.2 | NP_731267.1 | 2 | 1491 | 64 | |
| NM_169234.2 | NP_731268.1 | 2 | 1843 | 64 | ||
| GenBank mRNA | BT024234.1 | - | 2 | 1813 | 64 | |
More Information
Set Membership: |
|

hb
Genomic Map