Assay Details
| Gene Symbol: ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043227.2 | NP_001036692.1 | 3 | 747 | 136 | |
| NM_001043230.3 | NP_001036695.1 | 2 | 510 | 136 | ||
| NM_001202270.2 | NP_001189199.1 | 2 | 482 | 136 | ||
| NM_001202272.2 | NP_001189201.1 | 3 | 747 | 136 | ||
| NM_001202273.2 | NP_001189202.1 | 2 | 513 | 136 | ||
| NM_001202274.2 | NP_001189203.1 | 3 | 723 | 136 | ||
| NM_169280.4 | NP_731351.3 | 2 | 945 | 136 | ||
| NM_169281.4 | NP_731352.3 | 2 | 510 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001043227.2, NM_001043230.3, NM_001202270.2, NM_001202272.2, NM_001202273.2, NM_001202274.2, NM_169280.4, NM_169281.4 transcripts.
Set Membership: |
|

ps
Genomic Map