Assay Details
| Gene Symbol: Adk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300350.1 | NP_001287279.1 | 2 | 821 | 90 | |
| NM_001300351.1 | NP_001287280.1 | 2 | 521 | 90 | ||
| NM_079588.3 | NP_524312.1 | 2 | 828 | 90 | ||
| NM_169373.2 | NP_731529.1 | 2 | 514 | 90 | ||
| GenBank mRNA | AB050622.1 | - | 1 | 272 | 90 | |
| BT015244.1 | - | 2 | 828 | 90 | ||
More Information
Set Membership: |
|

Adk3
Genomic Map