Assay Details
| Gene Symbol: ATPsynE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202292.1 | NP_001189221.1 | 2 | 555 | 106 | |
| NM_142099.3 | NP_650356.1 | 2 | 434 | 106 | ||
| NM_169579.2 | NP_731903.1 | 2 | 490 | 106 | ||
| GenBank mRNA | AY060656.1 | - | 2 | 423 | 106 | |
More Information
Set Membership: |
|

ATPsynE
Genomic Map