Assay Details
| Gene Symbol: dco | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276203.1 | NP_001263132.1 | 4 | 2026 | 105 | |
| NM_001369991.1 | NP_001356928.1 | 3 | 2201 | 105 | ||
| NM_079863.3 | NP_524602.1 | 3 | 2638 | 105 | ||
| NM_170535.2 | NP_733414.1 | 3 | 1951 | 105 | ||
| NM_170536.3 | NP_733415.1 | 4 | 2033 | 105 | ||
| GenBank mRNA | AF132558.1 | - | 3 | 2201 | 105 | |
More Information
Set Membership: |
|

dco
Genomic Map