Assay Details
| Gene Symbol: Cpr31A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205956.2 | NP_995678.1 | 2 | 1059 | 60 | |
| GenBank mRNA | BT011451.1 | - | 2 | 1054 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr31A| Gene Symbol: Cpr31A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205956.2 | NP_995678.1 | 2 | 1059 | 60 | |
| GenBank mRNA | BT011451.1 | - | 2 | 1054 | 60 | |
Set Membership: |
|