Assay Details
| Gene Symbol: lectin-37Db | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014490.2 | NP_001014490.1 | 1 - 2 | 98 | 71 | |
| NM_001201918.1 | NP_001188847.1 | 1 - 2 | 98 | 71 | ||
| GenBank mRNA | KX531943.1 | - | 1 - 2 | 76 | 71 | |
More Information
Set Membership: |
|

lectin-37Db
Genomic Map