Assay Details
| Gene Symbol: Vkor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014533.2 | NP_001014533.1 | 2 - 3 | 255 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vkor| Gene Symbol: Vkor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014533.2 | NP_001014533.1 | 2 - 3 | 255 | 68 | |
Set Membership: |
|