Assay Details
| Gene Symbol: lectin-30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144378.4 | NP_652635.2 | 1 | 654 | 127 | |
| GenBank mRNA | BT032756.1 | - | 1 | 654 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lectin-30A| Gene Symbol: lectin-30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144378.4 | NP_652635.2 | 1 | 654 | 127 | |
| GenBank mRNA | BT032756.1 | - | 1 | 654 | 127 | |
Set Membership: |
|