Assay Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080483.3 | NP_536731.1 | 3 | 1856 | 80 | |
| GenBank mRNA | AY084154.1 | - | 3 | 1854 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cat| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080483.3 | NP_536731.1 | 3 | 1856 | 80 | |
| GenBank mRNA | AY084154.1 | - | 3 | 1854 | 80 | |
Set Membership: |
|