Assay Details
| Gene Symbol: Ald2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260385.1 | NP_001247314.1 | 1 | 532 | 122 | |
| NM_143219.2 | NP_651476.1 | 1 | 532 | 122 | ||
| GenBank mRNA | BT133270.1 | - | 1 | 532 | 122 | |
More Information
Set Membership: |
|

Ald2
Genomic Map