Assay Details
| Gene Symbol: Rab21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042856.3 | NP_001036321.2 | 2 | 971 | 163 | |
| NM_001042857.1 | NP_001036322.1 | 1 | 984 | 163 | ||
| NM_001272856.1 | NP_001259785.1 | 3 | 957 | 163 | ||
More Information
Set Membership: |
|

Rab21
Genomic Map