Assay Details
| Gene Symbol: Mst84Db | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079531.3 | NP_524255.2 | 2 | 244 | 72 | |
| GenBank mRNA | KX531777.1 | - | 1 | 175 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mst84Db| Gene Symbol: Mst84Db | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079531.3 | NP_524255.2 | 2 | 244 | 72 | |
| GenBank mRNA | KX531777.1 | - | 1 | 175 | 72 | |
Set Membership: |
|