Assay Details
| Gene Symbol: Cpr47Ec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136813.2 | NP_610657.1 | 2 | 480 | 65 | |
| GenBank mRNA | BT100144.4 | - | 1 | 343 | 65 | |
| KX531948.1 | - | 1 | 343 | 65 | ||
More Information
Set Membership: |
|

Cpr47Ec
Genomic Map