Assay Details
| Gene Symbol: Naa80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014612.3 | NP_001014612.1 | 3 | 1372 | 145 | |
| NM_141685.5 | NP_649942.1 | 2 | 1278 | 145 | ||
| GenBank mRNA | AY060239.1 | - | 2 | 1258 | 145 | |
More Information
Set Membership: |
|

Naa80
Genomic Map