Assay Details
| Gene Symbol: Cpr67Fb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140163.2 | NP_648420.1 | 2 | 309 | 65 | |
| GenBank mRNA | KX531538.1 | - | 1 | 291 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr67Fb| Gene Symbol: Cpr67Fb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140163.2 | NP_648420.1 | 2 | 309 | 65 | |
| GenBank mRNA | KX531538.1 | - | 1 | 291 | 65 | |
Set Membership: |
|