Assay Details
Gene Symbol: DCX-EMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274921.1 | NP_001261850.1 | 1 | 1142 | 77 | |
NM_140471.3 | NP_648728.2 | 1 | 1142 | 77 | ||
GenBank mRNA | BT030411.1 | - | 1 | 738 | 77 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
