Assay Details
| Gene Symbol: Acp24A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042865.1 | NP_001036330.1 | 2 | 205 | 114 | |
| NM_001258928.2 | NP_001245857.1 | 2 | 221 | 114 | ||
| GenBank mRNA | BT022473.1 | - | 2 | 205 | 114 | |
| BT022485.1 | - | 2 | 205 | 114 | ||
| KX531542.1 | - | 2 | 176 | 114 | ||
More Information
Set Membership: |
|

Acp24A4
Genomic Map