Assay Details
| Gene Symbol: tyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031861.2 | NP_001027032.1 | 5 - 6 | 2058 | 72 | |
| NM_001038726.2 | NP_001033815.1 | 4 - 5 | 2017 | 72 | ||
| NM_001297798.1 | NP_001284727.1 | 5 - 6 | 2055 | 72 | ||
| NM_001297799.1 | NP_001284728.1 | 4 - 5 | 2252 | 72 | ||
| NM_001297801.1 | NP_001284730.1 | 5 - 6 | 2343 | 72 | ||
| NM_001382036.1 | NP_001368965.1 | 6 - 7 | 2622 | 72 | ||
| GenBank mRNA | AY119115.1 | - | 5 - 6 | 1960 | 72 | |
More Information
Set Membership: |
|

tyn
Genomic Map