Assay Details
| Gene Symbol: tyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031861.2 | NP_001027032.1 | 6 - 7 | 2362 | 66 | |
| NM_001038726.2 | NP_001033815.1 | 5 - 6 | 2321 | 66 | ||
| NM_001297798.1 | NP_001284727.1 | 6 - 7 | 2359 | 66 | ||
| NM_001297799.1 | NP_001284728.1 | 5 - 6 | 2556 | 66 | ||
| NM_001297801.1 | NP_001284730.1 | 6 - 7 | 2647 | 66 | ||
| NM_001382036.1 | NP_001368965.1 | 7 - 8 | 2926 | 66 | ||
| GenBank mRNA | AY119115.1 | - | 6 - 7 | 2264 | 66 | |
More Information
Set Membership: |
|

tyn
Genomic Map