Assay Details
| Gene Symbol: RhoGAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103385.2 | NP_001096855.1 | 12 - 13 | 4075 | 73 | |
| NM_001103386.2 | NP_001096856.1 | 12 - 13 | 4075 | 73 | ||
| NM_001297795.1 | NP_001284724.1 | 12 - 13 | 4076 | 73 | ||
| NM_001297796.1 | NP_001284725.1 | 12 - 13 | 4082 | 73 | ||
| GenBank mRNA | BT024449.1 | - | 12 - 13 | 4037 | 73 | |
More Information
Set Membership: |
|

RhoGAP1A
Genomic Map