Assay Details
| Gene Symbol: RhoGAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103385.2 | NP_001096855.1 | 3 - 4 | 1220 | 64 | |
| NM_001103386.2 | NP_001096856.1 | 3 - 4 | 1220 | 64 | ||
| NM_001169155.1 | NP_001162626.1 | 2 - 3 | 1443 | 64 | ||
| NM_001297795.1 | NP_001284724.1 | 3 - 4 | 1224 | 64 | ||
| NM_001297796.1 | NP_001284725.1 | 3 - 4 | 1224 | 64 | ||
| NM_001297797.1 | NP_001284726.1 | 2 - 3 | 1013 | 64 | ||
| GenBank mRNA | BT024449.1 | - | 3 - 4 | 1182 | 64 | |
| BT081989.1 | - | 2 - 3 | 1443 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT081989.1, NM_001169155.1, NM_001297797.1 transcripts.
Set Membership: |
|

RhoGAP1A
Genomic Map