Assay Details
| Gene Symbol: CG17528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042997.2 | NP_001036462.1 | 5 - 6 | 2387 | 117 | |
| NM_001042998.2 | NP_001036463.1 | 5 - 6 | 2181 | 117 | ||
More Information
Set Membership: |
|

CG17528
Genomic Map