Assay Details
| Gene Symbol: stnB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042853.3 | NP_001036318.2 | 5 - 6 | 5677 | 100 | |
| NM_001042854.3 | NP_001036319.2 | 5 - 6 | 5803 | 100 | ||
| NM_001298610.1 | NP_001285539.1 | 5 - 6 | 5865 | 100 | ||
| NM_001298611.1 | NP_001285540.1 | 6 - 7 | 5740 | 100 | ||
| Gene Symbol: stnA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042851.3 | NP_001036316.1 | 5 - 6 | 5677 | 100 | |
| NM_001042852.3 | NP_001036317.1 | 5 - 6 | 5803 | 100 | ||
| NM_001298608.1 | NP_001285537.1 | 5 - 6 | 5865 | 100 | ||
| NM_001298609.1 | NP_001285538.1 | 6 - 7 | 5740 | 100 | ||
More Information
Set Membership: |
|

stnB
Genomic Map