Assay Details
| Gene Symbol: stnA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042851.3 | NP_001036316.1 | 6 - 7 | 5917 | 72 | |
| NM_001042852.3 | NP_001036317.1 | 6 - 7 | 6043 | 72 | ||
| NM_001298608.1 | NP_001285537.1 | 6 - 7 | 6105 | 72 | ||
| NM_001298609.1 | NP_001285538.1 | 7 - 8 | 5980 | 72 | ||
| GenBank mRNA | BT011172.1 | - | 6 - 7 | 5917 | 72 | |
| Gene Symbol: stnB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042853.3 | NP_001036318.2 | 6 - 7 | 5917 | 72 | |
| NM_001042854.3 | NP_001036319.2 | 6 - 7 | 6043 | 72 | ||
| NM_001298610.1 | NP_001285539.1 | 6 - 7 | 6105 | 72 | ||
| NM_001298611.1 | NP_001285540.1 | 7 - 8 | 5980 | 72 | ||
More Information
Set Membership: |
|

stnA
Genomic Map