Assay Details
| Gene Symbol: solo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032103.2 | NP_001027274.2 | 4 - 5 | 1624 | 75 | |
| NM_001316394.1 | NP_001303323.1 | 4 - 5 | 1578 | 75 | ||
| GenBank mRNA | BT099809.1 | - | 4 - 5 | 1575 | 75 | |
| DQ851162.1 | - | 4 - 5 | 1562 | 75 | ||
More Information
Set Membership: |
|

solo
Genomic Map