Assay Details
| Gene Symbol: CG33978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038712.3 | NP_001033801.3 | 3 - 4 | 273 | 85 | |
| NM_001258478.1 | NP_001245407.1 | 3 - 4 | 360 | 85 | ||
More Information
Set Membership: |
|

CG33978
Genomic Map