Assay Details
| Gene Symbol: CG33978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272115.1 | NP_001259044.1 | 5 - 6 | 908 | 103 | |
| NM_001272116.1 | NP_001259045.1 | 5 - 6 | 815 | 103 | ||
| NM_001272117.1 | NP_001259046.1 | 5 - 6 | 815 | 103 | ||
More Information
Set Membership: |
|

CG33978
Genomic Map