Assay Details
| Gene Symbol: fab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137425.4 | NP_611269.1 | 4 | 2990 | 106 | |
| NM_206158.3 | NP_995880.1 | 4 | 2990 | 106 | ||
| GenBank mRNA | BT099666.1 | - | 4 | 2925 | 106 | |
| Gene Symbol: CG33981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038865.3 | NP_001033954.1 | 4 | 2990 | 106 | |
| NM_001038866.3 | NP_001033955.1 | 4 | 2990 | 106 | ||
| NM_001274133.2 | NP_001261062.1 | 4 | 2990 | 106 | ||
More Information
Set Membership: |
|

fab1
Genomic Map