Assay Details
| Gene Symbol: Pax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165296.3 | NP_724186.2 | 3 | 777 | 54 | |
| GenBank mRNA | AB056465.1 | - | 3 | 761 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pax| Gene Symbol: Pax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165296.3 | NP_724186.2 | 3 | 777 | 54 | |
| GenBank mRNA | AB056465.1 | - | 3 | 761 | 54 | |
Set Membership: |
|