Assay Details
| Gene Symbol: CG17691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015354.4 | NP_001015354.3 | 4 - 5 | 861 | 129 | |
| NM_001110548.2 | NP_001104018.1 | 5 - 6 | 884 | 129 | ||
More Information
Set Membership: |
|

CG17691
Genomic Map